Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026307.7 | NP_001021478.1 | 4 - 5 | 542 | 136 | |
NM_001383463.1 | 3 - 4 | 210 | 138 | |||
GenBank mRNA | AF440800.2 | - | 3 - 4 | 222 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |