Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129034.3 | NP_001122506.1 | 20 - 21 | 4482 | 97 | |
NM_001136316.4 | NP_001129788.1 | 14 - 15 | 3447 | 97 | ||
NM_001373178.2 | NP_001359707.1 | 22 - 23 | 4633 | 97 | ||
NM_059914.7 | NP_492315.4 | 23 - 24 | 4706 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |