Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sig-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373191.2 | NP_001359651.1 | 3 - 4 | 427 | 88 | |
NM_059942.8 | NP_492343.2 | 3 - 4 | 563 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |