Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059980.6 | NP_492381.3 | 2 - 3 | 221 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |