Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060043.5 | NP_492444.4 | 5 - 6 | 656 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |