Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026238.5 | NP_001021409.2 | 3 - 4 | 537 | 93 | |
NM_001030057.4 | NP_001025228.2 | 5 - 6 | 1117 | 93 | ||
NM_001313465.3 | NP_001300394.1 | 10 | 1283 | 93 | ||
NM_001313466.5 | NP_001300395.1 | 11 | 1373 | 93 | ||
NM_001377616.3 | NP_001364651.1 | 8 - 9 | 962 | 93 | ||
NM_001377617.3 | NP_001364650.1 | 5 - 6 | 686 | 93 | ||
GenBank mRNA | EF443133.1 | - | 8 - 9 | 962 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |