Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060107.7 | NP_492508.1 | 1 - 2 | 312 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |