Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ahr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025865.4 | NP_001021036.1 | 4 - 5 | 655 | 77 | |
NM_001025866.5 | NP_001021037.1 | 3 - 4 | 589 | 77 | ||
GenBank mRNA | AF039570.1 | - | 4 - 5 | 651 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |