Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025808.5 | NP_001020979.1 | 2 - 3 | 377 | 90 | |
NM_001025809.2 | NP_001020980.1 | 2 - 3 | 319 | 90 | ||
GenBank mRNA | AF003925.1 | - | 2 - 3 | 224 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |