Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060214.5 | NP_492615.2 | 7 - 8 | 968 | 67 | |
GenBank mRNA | AY204178.1 | - | 7 - 8 | 988 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |