Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264316.2 | NP_001251245.1 | 6 - 7 | 1212 | 84 | |
NM_001264317.3 | NP_001251246.1 | 5 - 6 | 903 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |