Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mut-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060259.6 | NP_492660.3 | 7 - 8 | 1298 | 127 | |
NM_060260.9 | NP_492661.3 | 7 - 8 | 1286 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |