Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381229.1 | NP_001367884.1 | 3 - 4 | 309 | 77 | |
NM_058280.9 | NP_490681.2 | 3 - 4 | 310 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |