Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306270.3 | NP_001293199.1 | 1 - 2 | 207 | 68 | |
NM_058422.7 | NP_490823.1 | 3 - 4 | 497 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |