Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tub-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058512.5 | NP_490913.2 | 2 - 3 | 1537 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |