Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058686.5 | NP_491087.1 | 2 - 3 | 269 | 75 | |
GenBank mRNA | U21107.1 | - | 2 - 3 | 288 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |