Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025843.3 | NP_001021014.1 | 5 - 6 | 1293 | 109 | |
NM_001025845.4 | NP_001021016.1 | 5 - 6 | 1293 | 109 | ||
GenBank mRNA | AF144258.1 | - | 5 - 6 | 1260 | 109 | |
AF144259.1 | - | 5 - 6 | 1260 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |