Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383031.1 | 11 - 12 | 2884 | 113 | ||
GenBank mRNA | AF386744.1 | - | 6 - 7 | 1038 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |