Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dom-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059095.7 | NP_491496.1 | 3 - 4 | 356 | 84 | |
GenBank mRNA | U34893.1 | - | 2 - 3 | 255 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |