Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfe-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025878.7 | NP_001021049.1 | 3 - 4 | 196 | 68 | |
NM_001262849.3 | NP_001249778.1 | 3 - 4 | 179 | 68 | ||
NM_001372686.1 | NP_001359911.1 | 2 - 3 | 118 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |