Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059157.7 | NP_491558.1 | 4 - 5 | 367 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |