Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380377.2 | NP_001367356.1 | 2 - 3 | 182 | 85 | |
NM_059282.5 | NP_491683.1 | 3 - 4 | 234 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |