Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171392.7 | NP_741457.2 | 2 - 3 | 202 | 65 | |
NM_171913.6 | NP_741458.2 | 2 - 3 | 191 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |