Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171475.5 | NP_741557.2 | 10 - 11 | 1893 | 85 | |
NM_171476.5 | NP_741558.2 | 10 - 11 | 1893 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |