Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171516.3 | NP_741602.1 | 4 - 5 | 500 | 98 | |
GenBank mRNA | AF216966.1 | - | 4 - 5 | 500 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |