Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028662.4 | NP_001023833.1 | 6 - 7 | 1218 | 70 | |
NM_001047640.4 | NP_001041105.1 | 7 - 8 | 1760 | 70 | ||
NM_001373098.1 | NP_001359645.1 | 8 - 9 | 1901 | 70 | ||
NM_171533.6 | NP_741621.1 | 9 - 10 | 1996 | 70 | ||
GenBank mRNA | AF178536.1 | - | 9 - 10 | 1998 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |