Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171550.5 | NP_741638.1 | 5 - 6 | 882 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |