Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pro-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171594.5 | NP_741684.2 | 1 - 2 | 195 | 119 | |
GenBank mRNA | EU068466.1 | - | 1 - 2 | 187 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |