Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171615.7 | NP_741707.3 | 8 - 9 | 2378 | 131 | |
NM_171616.5 | NP_741708.1 | 8 - 9 | 1262 | 131 | ||
NM_171617.5 | NP_741709.1 | 7 - 8 | 1402 | 131 | ||
GenBank mRNA | AF456360.2 | - | 8 - 9 | 1286 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |