Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348597.2 | NP_001335515.1 | 19 - 20 | 4456 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |