Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270068.6 | NP_001256997.2 | 2 - 3 | 385 | 63 | |
NM_001313456.4 | NP_001300385.1 | 2 | 367 | 63 | ||
NM_171645.6 | NP_741740.3 | 2 - 3 | 384 | 63 | ||
NM_171646.5 | NP_741741.2 | 2 - 3 | 386 | 63 | ||
GenBank mRNA | EU729334.1 | - | 2 - 3 | 367 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |