Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029819.7 | NP_001024990.1 | 11 - 12 | 3828 | 83 | |
NM_171652.7 | NP_741748.2 | 11 - 12 | 3814 | 83 | ||
GenBank mRNA | AF041053.1 | - | 11 - 12 | 3743 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |