Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171662.9 | NP_741761.1 | 6 - 7 | 2052 | 52 | |
NM_171663.4 | NP_741762.1 | 6 - 7 | 1816 | 52 | ||
NM_171664.7 | NP_741763.1 | 5 - 6 | 1658 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |