Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 13 - 14 | 1447 | 91 | |
NM_001373290.4 | NP_001360675.1 | 10 - 11 | 1158 | 91 | ||
NM_171949.8 | NP_741779.1 | 11 - 12 | 1321 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |