Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 3 - 4 | 289 | 105 | |
NM_171949.8 | NP_741779.1 | 1 - 2 | 163 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |