Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 6 - 7 | 590 | 98 | |
NM_001373290.4 | NP_001360675.1 | 3 - 4 | 301 | 98 | ||
NM_171949.8 | NP_741779.1 | 4 - 5 | 464 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |