Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029732.3 | NP_001024903.1 | 3 - 4 | 456 | 57 | |
NM_001375044.2 | NP_001362132.1 | 2 - 3 | 393 | 57 | ||
NM_171699.6 | NP_741807.2 | 4 - 5 | 562 | 57 | ||
GenBank mRNA | AF407572.1 | - | 3 - 4 | 456 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |