Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383608.1 | 1 - 2 | 219 | 89 | ||
NM_171725.8 | NP_741836.1 | 2 - 3 | 465 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |