Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029346.4 | NP_001024517.1 | 12 - 13 | 1919 | 84 | |
NM_001373417.3 | NP_001360728.1 | 13 | 2025 | 84 | ||
NM_001373419.3 | NP_001359883.1 | 13 - 14 | 2103 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |