Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171790.7 | NP_741917.1 | 2 - 3 | 240 | 117 | |
GenBank mRNA | U93195.1 | - | 2 - 3 | 238 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |