Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027681.8 | NP_001022852.1 | 5 - 6 | 909 | 78 | |
NM_001379977.1 | NP_001366945.1 | 6 - 7 | 987 | 78 | ||
NM_001383020.1 | 5 - 6 | 903 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |