Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171897.7 | NP_741299.2 | 7 - 8 | 1375 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |