Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zen-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028138.2 | NP_001023309.1 | 4 - 5 | 1249 | 106 | |
NM_001028139.3 | NP_001023310.1 | 4 - 5 | 1245 | 106 | ||
NM_001028140.5 | NP_001023311.1 | 4 - 5 | 1255 | 106 | ||
NM_001372836.1 | NP_001360082.1 | 4 - 5 | 1255 | 106 | ||
NM_001372837.3 | NP_001360296.1 | 4 - 5 | 1249 | 106 | ||
NM_001372838.2 | NP_001360295.1 | 4 - 5 | 1253 | 106 | ||
GenBank mRNA | AF057567.1 | - | 4 - 5 | 1240 | 106 | |
AF057568.1 | - | 4 - 5 | 1240 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |