Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307092.4 | NP_001294021.1 | 3 - 4 | 209 | 80 | |
NM_001380456.1 | NP_001367187.1 | 1 - 2 | 55 | 80 | ||
NM_001380457.2 | NP_001367146.1 | 2 - 3 | 154 | 80 | ||
NM_171919.7 | NP_741490.1 | 3 - 4 | 209 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |