Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 5 - 6 | 482 | 73 | |
NM_171949.8 | NP_741779.1 | 3 - 4 | 356 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |