Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066742.5 | NP_499143.1 | 4 - 5 | 642 | 112 | |
NM_181940.6 | NP_871669.1 | 4 - 5 | 644 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |