Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381396.1 | NP_001367814.1 | 3 - 4 | 716 | 115 | |
NM_001381397.2 | NP_001367813.1 | 4 - 5 | 788 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |