Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lips-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383781.1 | 3 - 4 | 1397 | 89 | ||
NM_061355.4 | NP_493756.1 | 4 - 5 | 616 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |