Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368589.3 | NP_001355398.1 | 6 - 7 | 555 | 123 | |
NM_001368590.4 | NP_001355414.1 | 3857 | 123 | |||
NM_001381447.1 | NP_001367591.1 | 5 - 6 | 593 | 123 | ||
NM_001381448.2 | NP_001367590.1 | 5 - 6 | 604 | 123 | ||
NM_062827.5 | NP_495228.1 | 4 - 5 | 851 | 123 | ||
GenBank mRNA | AY887905.1 | - | 4 - 5 | 873 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |