Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182311.4 | NP_872111.2 | 4 - 5 | 829 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |